Learn More
Medchemexpress LLC Delphinidin chloride | 528-53-0 | MFCD00016663 | 5 MG
SDP
Catalog No. 5000420432
Encompass_Preferred
Delphinidin chloride is an anthocyanin isolated from berries and red wine. It demonstrates endothelium-dependent vasodilation and anticancer effects, modulating JAK/STAT3 and MAPK signaling to induce apoptosis in HCT116 cells. It is also a potent inhibitor of EGFR, halting downstream signaling cascades.
  • Isolated from berries and red wine.
  • Exhibits endothelium-dependent vasodilation.
  • Demonstrates anticancer activity.
  • Modulates JAK/STAT3 and MAPK signaling.
  • Induces apoptosis in HCT116 cells.
  • Potent inhibitor of EGFR (IC50: 1.3 μM).
Catalog No. 50-004-20432 Supplier Medchemexpress LLC Supplier No. HYN24095MG
Delphinidin chloride is an anthocyanin isolated from berries and red wine. It demonstrates endothelium-dependent vasodilation and anticancer effects, modulating JAK/STAT3 and MAPK signaling to induce apoptosis in HCT116 cells. It is also a potent inhibitor of EGFR, halting downstream signaling cascades.
  • Isolated from berries and red wine.
  • Exhibits endothelium-dependent vasodilation.
  • Demonstrates anticancer activity.
  • Modulates JAK/STAT3 and MAPK signaling.
  • Induces apoptosis in HCT116 cells.
  • Potent inhibitor of EGFR (IC50: 1.3 μM).