Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Delphinidin chloride, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014470 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4470 1 mg
11-101-4469 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4470 Supplier MilliporeSigma Supelco Supplier No. 437251MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delphinidin chloride is commonly found in delphinium flowers. It can be obtained, via hydrolysis of delphinin chloride.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 528-53-0
Physical Form Neat
Linear Formula C15H11 ClO7
MDL Number MFCD00016663
Synonym 3,3 ′,4 ′,5,5 ′,7-Hexahydroxyflavylium chloride; 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-1-benzopyrylium chloride
Type Analytical Standard
Quantity 1 mg
Formula Weight 338.70
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.