Learn More
Cayman Chemical DelphInIdIn3OrutInosIdec 10mg
SDP
Catalog No. 502946836
Encompass_Preferred
A glycosylated anthocyanin with antioxidant activity; inhibits TBHP-induced loss of viability in isolated rat aortic SMCs, but not MH1C1 cells, at 100 µM; prevents TBHP-induced DNA damage, as well as inhibits TBHP-induced increases in MDA levels, in SMCs; displays a hydrogen peroxide-induced and pH-dependent chemiluminescence peak at pH 7
Catalog No. 50-294-6836 Supplier Cayman Chemical Supplier No. 3355210 MG
A glycosylated anthocyanin with antioxidant activity; inhibits TBHP-induced loss of viability in isolated rat aortic SMCs, but not MH1C1 cells, at 100 µM; prevents TBHP-induced DNA damage, as well as inhibits TBHP-induced increases in MDA levels, in SMCs; displays a hydrogen peroxide-induced and pH-dependent chemiluminescence peak at pH 7