Learn More
Analytical Sales & Services Delrin Column Coupler
Catalog No. 5000806482
Encompass
Advantage PEEK Column Couplers Includes Two Pair of Two-Piece Fingertight Fittings and One Length of PEEK 0.007: ID Tubing
Qty/Pkg: EACH
Catalog No. 50-008-06482 Supplier Analytical Sales & Services Supplier No. 64284
Add to Cart
Add to Cart
Advantage PEEK Column Couplers Includes Two Pair of Two-Piece Fingertight Fittings and One Length of PEEK 0.007: ID Tubing
Qty/Pkg: EACH