Learn More
Analytical Sales & Services Delrin Flangeless Nut for 1/16" OD Tubing
Catalog No. 5000804600
Encompass
Delrin 1/4-28 Flangeless Male Nut for 1/16" OD Tubing Cream
Qty/Pkg: EACH
Catalog No. 50-008-04600 Supplier Analytical Sales & Services Supplier No. 59204
Delrin 1/4-28 Flangeless Male Nut for 1/16" OD Tubing Cream
Qty/Pkg: EACH