Learn More
Analytical Sales & Services Delrin Flangeless Nut for 1/16" OD Tubing
Catalog No. 5000804667
Encompass
Blue Delrin Nuts • Rated to 2500 psi on PEEKª tubing • Nuts are compatible with all 1/4-28 fitting systems
Catalog No. 50-008-04667 Supplier Analytical Sales & Services Supplier No. 59206X
Add to Cart
Add to Cart
Blue Delrin Nuts • Rated to 2500 psi on PEEKª tubing • Nuts are compatible with all 1/4-28 fitting systems