Learn More
Analytical Sales & Services Delrin Flangeless Nut for 1/16" OD Tubing
Catalog No. 5000804761
Encompass
Delrin 1/4-28 Flangeless Male Nut for 1/16" OD Tubing Black
Qty/Pkg: 10PK
Catalog No. 50-008-04761 Supplier Analytical Sales & Services Supplier No. 59201X
Delrin 1/4-28 Flangeless Male Nut for 1/16" OD Tubing Black
Qty/Pkg: 10PK