Learn More
Analytical Sales & Services Delrin Flangeless Nut for 1/16" OD Tubing
Catalog No. 5000804851
Encompass
Delrin 1/4-28 Flangeless Male Nut for 1/16" OD Tubing Cream
Qty/Pkg: 10PK
Catalog No. 50-008-04851 Supplier Analytical Sales & Services Supplier No. 59204X
Delrin 1/4-28 Flangeless Male Nut for 1/16" OD Tubing Cream
Qty/Pkg: 10PK