Learn More
World Precision Instrument Delrin Plastic Tweezers, 11.8cm, Curved
SDP
Catalog No. 502539490
Encompass
Delrin plastic is an engineering thermoplastic used in precision parts requiring high stiffness, low friction and excellent dimensional stability. As with many other synthetic polymers, it is produced by different chemical firms with slightly different formulas and sold variously by such names as Delrin, Celcon, Duracon and Hostaform.
Catalog No. 50-253-9490 Supplier World Precision Instrument Supplier No. 503362
Delrin plastic is an engineering thermoplastic used in precision parts requiring high stiffness, low friction and excellent dimensional stability. As with many other synthetic polymers, it is produced by different chemical firms with slightly different formulas and sold variously by such names as Delrin, Celcon, Duracon and Hostaform.