Learn More
Analytical Sales & Services Delrin Short Flangeless Nut for 1/8" OD Tubing
Catalog No. 5000804872
Encompass
Black Delrin Short Flangeless Male Nut for 1/8" OD Tubing 1/4-28
Qty/Pkg: 10PK
Catalog No. 50-008-04872 Supplier Analytical Sales & Services Supplier No. 59308X
Add to Cart
Add to Cart
Black Delrin Short Flangeless Male Nut for 1/8" OD Tubing 1/4-28
Qty/Pkg: 10PK