Learn More
Analytical Sales & Services Delrin Slip Style Adapter - Male Luer to 1/4-28 Male
Catalog No. 5000805078
Encompass
Delrin Slip Style Luer Adapter Male Luer to 1/4-28 Male
Qty/Pkg: EACH
Catalog No. 50-008-05078 Supplier Analytical Sales & Services Supplier No. 59605
Delrin Slip Style Luer Adapter Male Luer to 1/4-28 Male
Qty/Pkg: EACH