Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Delsoline | 509-18-2 | MFCD00274545 | 99.1% | 467.6 | C25H41NO7 | 10 MG
SDP

Catalog No. 5000296402
Encompass_Preferred

Delsoline is a naturally occurring alkaloid isolated from Delphinium anthriscifolium Hance. It exhibits curare-like and ganglion-blocking activity and is supplied as a research reagent for studies of neuromuscular pharmacology and biochemical effects. Typical attributes include high purity and a defined molecular weight.

  • High purity (99.06%).
  • Molecular formula C25H41NO7.
  • Molecular weight 467.6 g/mol.
  • Supplied as a 10 mg solid.
  • Used for neuromuscular and ganglionic activity studies.

Catalog No. 50-002-96402 Supplier Medchemexpress LLC Supplier No. HYN078910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delsoline is a naturally occurring alkaloid isolated from Delphinium anthriscifolium Hance. It exhibits curare-like and ganglion-blocking activity and is supplied as a research reagent for studies of neuromuscular pharmacology and biochemical effects. Typical attributes include high purity and a defined molecular weight.

  • High purity (99.06%).
  • Molecular formula C25H41NO7.
  • Molecular weight 467.6 g/mol.
  • Supplied as a 10 mg solid.
  • Used for neuromuscular and ganglionic activity studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.