Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Octanolactone 98.0+%, TCI America™
SDP

Catalog No. O022025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O022025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O022025G Supplier TCI America Supplier No. O022025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 698-76-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00144051
InChI Key FYTRVXSHONWYNE-UHFFFAOYSA-N
Synonym delta-octanolactone, delta-octalactone, 5-octanolide, 5-hydroxyoctanoic acid lactone, 2h-pyran-2-one, tetrahydro-6-propyl, 5-octalactone, .delta.-octalactone, tetrahydro-6-propyl-2h-pyran-2-one, delta-octanolide, fema no. 3214
PubChem CID 12777
IUPAC Name 6-propyloxan-2-one
SMILES CCCC1CCCC(=O)O1

Specifications

Melting Point -14°C
Color Yellow
Boiling Point 238°C
Quantity 25 g
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material delta-Octanolactone
RTECSNumber UQ1355500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.