Learn More
Waters Corp Waters Column, C18, 100Å, 15 μm, 40 mm x 100 mm
Catalog No. 50785433 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These packings are designed for the separation of peptides, proteins, and natural products. They are based on a stable, bonded, endcapped spherical silica, offering consistent and predictable scale-up capabilities for purification.
  • Ideal for the separation of peptides, proteins, and natural products.
  • Based on highly stable, bonded, endcapped 15 μm spherical silica.
  • Features a 100Å pore size with C18 bonded phase.
  • Provides consistent and predictable scale-up capabilities for milligram-to-gram scale purification.
  • The same chemistry is available on a high-efficiency 5 μm particle, allowing for purity checks of fractions.
  • Format is a Radial-Pak for preparative chromatography.
Catalog No. 50-785-433 Supplier Waters Corp Supplier No. WAT037688
These packings are designed for the separation of peptides, proteins, and natural products. They are based on a stable, bonded, endcapped spherical silica, offering consistent and predictable scale-up capabilities for purification.
  • Ideal for the separation of peptides, proteins, and natural products.
  • Based on highly stable, bonded, endcapped 15 μm spherical silica.
  • Features a 100Å pore size with C18 bonded phase.
  • Provides consistent and predictable scale-up capabilities for milligram-to-gram scale purification.
  • The same chemistry is available on a high-efficiency 5 μm particle, allowing for purity checks of fractions.
  • Format is a Radial-Pak for preparative chromatography.