Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Radial-Pak Column, C18, 15 μm, 100 Å

Catalog No. 50785077 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These Radial-Pak columns are engineered for the separation of peptides, proteins, and natural products. They are built upon highly stable, bonded, endcapped spherical silica packings and are designed to offer consistent and predictable scale-up capabilities for purification from milligram to gram scale. The consistent chemistry across various particle sizes facilitates easy purity checks of fractions.

  • 15 μm spherical silica particles.
  • C18 chemistry with 100 Å pore size.
  • pH range of 2 to 8.
  • Maximum temperature of 45 °C.
  • Maximum pressure of 600 psi (40 Bar).
  • Endcapped with low silanol activity.
  • Molecular weight range of 1000 to 15000.
  • Dimensions: 47 mm inner diameter, 300 mm length.
  • 17% carbon load.
  • Suitable for HPLC preparative systems, LC/MS, and LC techniques.

Catalog No. 50-785-077 Supplier Waters Corp Supplier No. WAT015401
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These Radial-Pak columns are engineered for the separation of peptides, proteins, and natural products. They are built upon highly stable, bonded, endcapped spherical silica packings and are designed to offer consistent and predictable scale-up capabilities for purification from milligram to gram scale. The consistent chemistry across various particle sizes facilitates easy purity checks of fractions.

  • 15 μm spherical silica particles.
  • C18 chemistry with 100 Å pore size.
  • pH range of 2 to 8.
  • Maximum temperature of 45 °C.
  • Maximum pressure of 600 psi (40 Bar).
  • Endcapped with low silanol activity.
  • Molecular weight range of 1000 to 15000.
  • Dimensions: 47 mm inner diameter, 300 mm length.
  • 17% carbon load.
  • Suitable for HPLC preparative systems, LC/MS, and LC techniques.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.