Learn More
Waters Corp Radial-Pak Cartridge, C18, 15 μm, 300Å, 47 mm x 300 mm
Catalog No. 50784998 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Delta-Pak packings are designed for the separation of peptides, proteins, and natural products. They are based on highly stable, bonded, endcapped 5μm or 15μm spherical silica. These packings are available in 100Å and 300Å pore sizes with C4 or C18 bonded phases, offered in various cartridge and column configurations. This provides consistent and predictable scale-up for milligram-to-gram scale purification, and the same chemistry on a high-efficiency 5μm particle allows for easy purity checks of fractions.
  • Separates peptides, proteins, and natural products.
  • Based on highly stable, bonded, endcapped 5μm or 15μm spherical silica.
  • Available in 100Å and 300Å pore sizes with C4 or C18 bonded phases.
  • Consistent and predictable scale-up capabilities for milligram-to-gram purification.
  • High-efficiency 5μm particle enables easy purity checks of fractions.
Catalog No. 50-784-998 Supplier Waters Corp Supplier No. WAT010988
Delta-Pak packings are designed for the separation of peptides, proteins, and natural products. They are based on highly stable, bonded, endcapped 5μm or 15μm spherical silica. These packings are available in 100Å and 300Å pore sizes with C4 or C18 bonded phases, offered in various cartridge and column configurations. This provides consistent and predictable scale-up for milligram-to-gram scale purification, and the same chemistry on a high-efficiency 5μm particle allows for easy purity checks of fractions.
  • Separates peptides, proteins, and natural products.
  • Based on highly stable, bonded, endcapped 5μm or 15μm spherical silica.
  • Available in 100Å and 300Å pore sizes with C4 or C18 bonded phases.
  • Consistent and predictable scale-up capabilities for milligram-to-gram purification.
  • High-efficiency 5μm particle enables easy purity checks of fractions.