Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Delta-Pak C18 Radial-Pak Column, 300Å, 15 μm, 25 mm x 100 mm

Catalog No. 50785456 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Waters Delta-Pak columns are designed for reversed-phase separation of proteins or peptides. Utilizing silica-based, reversed-phase spherical particles, these Radial Compression Cartridges (Radial-Paks™) offer predictable scale-up capabilities for purification from analytical to milligram-to-gram scales.

  • Silica-based, reversed-phase spherical particles with C4 or C18 ligands and end-capping.
  • Available in 5 μm or 15 μm particle sizes.
  • Features 100 Å or 300 Å pore sizes for reversed-phase separation.
  • Ideal for reversed-phase separation of proteins or peptides.
  • Recommended operational pH range: 2 to 8.
  • Tolerates pressures up to 6,000 psi (400 bar or 40 Mpa); avoid >4,000 to 5,000 psi for longevity.
  • Optimal operating temperatures: 20°C to 45°C for enhanced selectivity and mass transfer.

Catalog No. 50-785-456 Supplier Waters Corp Supplier No. WAT038507
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waters Delta-Pak columns are designed for reversed-phase separation of proteins or peptides. Utilizing silica-based, reversed-phase spherical particles, these Radial Compression Cartridges (Radial-Paks™) offer predictable scale-up capabilities for purification from analytical to milligram-to-gram scales.

  • Silica-based, reversed-phase spherical particles with C4 or C18 ligands and end-capping.
  • Available in 5 μm or 15 μm particle sizes.
  • Features 100 Å or 300 Å pore sizes for reversed-phase separation.
  • Ideal for reversed-phase separation of proteins or peptides.
  • Recommended operational pH range: 2 to 8.
  • Tolerates pressures up to 6,000 psi (400 bar or 40 Mpa); avoid >4,000 to 5,000 psi for longevity.
  • Optimal operating temperatures: 20°C to 45°C for enhanced selectivity and mass transfer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.