Learn More
Waters Corp Waters C18 Column, 15 μm, 100 Å, 3.9 mm × 300 mm
Catalog No. 50785055 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C18 column is primarily used for legacy methods. It features a highly stable, bonded, endcapped 15 μm spherical silica packing, available in various cartridge and column configurations. It provides consistent and predictable scale-up capabilities for milligram-to-gram scale purification, and the availability of the same chemistry on a high-efficiency 5 μm particle allows for easy checking of fraction purity.
  • Based on a highly stable, bonded, endcapped 15 μm spherical silica.
  • Features 100 Å pore size and C18 bonded phase.
  • Available in different cartridge and column configurations.
  • Provides consistent and predictable scale-up capabilities for milligram-to-gram scale purification.
  • Allows for easy purity checks of fractions when the same chemistry is available on a high-efficiency 5 μm particle.
Catalog No. 50-785-055 Supplier Waters Corp Supplier No. WAT011797
This C18 column is primarily used for legacy methods. It features a highly stable, bonded, endcapped 15 μm spherical silica packing, available in various cartridge and column configurations. It provides consistent and predictable scale-up capabilities for milligram-to-gram scale purification, and the availability of the same chemistry on a high-efficiency 5 μm particle allows for easy checking of fraction purity.
  • Based on a highly stable, bonded, endcapped 15 μm spherical silica.
  • Features 100 Å pore size and C18 bonded phase.
  • Available in different cartridge and column configurations.
  • Provides consistent and predictable scale-up capabilities for milligram-to-gram scale purification.
  • Allows for easy purity checks of fractions when the same chemistry is available on a high-efficiency 5 μm particle.