Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Prep Column, C18, 15 μm, 300 Å Pore, 19 mm x 300 mm

Catalog No. 50785062 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Delta-Pak C18 Prep Columns are designed for reversed-phase separation in HPLC preparatory systems. Built on highly stable, bonded, endcapped spherical silica, these columns feature a 15 μm particle size and 300 Å pore size, offering consistent scale-up capabilities for purification from milligrams to grams.

  • C18 chemistry for reversed-phase separation.
  • Constructed with spherical silica particles.
  • 15 μm particle size.
  • 300 Å pore size.
  • Preparative column format.
  • Endcapped for enhanced performance.
  • pH range of 2 to 8.
  • Maximum operating temperature of 45 °C.
  • Maximum pressure of 6000 psi (415 bar).
  • 19 mm inner diameter.
  • 300 mm length.
  • Low silanol activity.
  • Molecular weight range of 1000 to 30000.
  • Suitable for LC/MS and LC techniques.
  • USP classification L1.

Catalog No. 50-785-062 Supplier Waters Corp Supplier No. WAT011804
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Delta-Pak C18 Prep Columns are designed for reversed-phase separation in HPLC preparatory systems. Built on highly stable, bonded, endcapped spherical silica, these columns feature a 15 μm particle size and 300 Å pore size, offering consistent scale-up capabilities for purification from milligrams to grams.

  • C18 chemistry for reversed-phase separation.
  • Constructed with spherical silica particles.
  • 15 μm particle size.
  • 300 Å pore size.
  • Preparative column format.
  • Endcapped for enhanced performance.
  • pH range of 2 to 8.
  • Maximum operating temperature of 45 °C.
  • Maximum pressure of 6000 psi (415 bar).
  • 19 mm inner diameter.
  • 300 mm length.
  • Low silanol activity.
  • Molecular weight range of 1000 to 30000.
  • Suitable for LC/MS and LC techniques.
  • USP classification L1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.