Learn More
Waters Corp DELTA-PAK C18 Column, 5 μm, 300 Å, 3.9 x 300 mm
Catalog No. 50785053 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The DELTA-PAK C18 Column is designed for high-efficiency separations. It features 5 μm, 300 Å Delta-Pak C18 bonded, spherical silica particles, making it suitable for reversed-phase separation applications.
  • Packed with 5 μm, 300 Å Delta-Pak C18 bonded, spherical silica particles.
  • High-efficiency, steel analysis column.
  • Used for the reversed-phase separation of proteins, peptides, and other biopolymers.
Catalog No. 50-785-053 Supplier Waters Corp Supplier No. WAT011795
The DELTA-PAK C18 Column is designed for high-efficiency separations. It features 5 μm, 300 Å Delta-Pak C18 bonded, spherical silica particles, making it suitable for reversed-phase separation applications.
  • Packed with 5 μm, 300 Å Delta-Pak C18 bonded, spherical silica particles.
  • High-efficiency, steel analysis column.
  • Used for the reversed-phase separation of proteins, peptides, and other biopolymers.