Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp HPLC Column, C18, 300 Å/5 μm, 2.1 x 150 mm

Catalog No. 50785198 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Delta-Pak HPLC column packing is designed for the separation of peptides, proteins, and natural products. It is based on a highly stable, bonded, endcapped spherical silica, offering consistent and predictable scale-up capabilities.

  • Pore size of 300 Å
  • Particle size of 5.0 μm
  • Length of 150 mm
  • Inner diameter of 2.1 mm
  • C18 (RP18, ODS, Octadecyl) reversed phase
  • pH range of 2 to 8
  • 17.0% carbon load
  • Endcapped
  • USP L1 number

Catalog No. 50-785-198 Supplier Waters Corp Supplier No. WAT023650
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This Delta-Pak HPLC column packing is designed for the separation of peptides, proteins, and natural products. It is based on a highly stable, bonded, endcapped spherical silica, offering consistent and predictable scale-up capabilities.

  • Pore size of 300 Å
  • Particle size of 5.0 μm
  • Length of 150 mm
  • Inner diameter of 2.1 mm
  • C18 (RP18, ODS, Octadecyl) reversed phase
  • pH range of 2 to 8
  • 17.0% carbon load
  • Endcapped
  • USP L1 number

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.