Learn More
Waters Corp Waters Corporation Guard Column, C18, 5 μm, 100 Å, 3.9 mm × 20 mm
Supplier: Waters Corp WAT046880
This C18 guard column features highly stable, bonded, and endcapped spherical silica particles with a 5 μm particle size and 100 Å pore size. Designed for reversed phase separation, it operates within a pH range of 2 to 8 and can withstand pressures up to 6000 psi (415 Bar). It is primarily used for legacy methods and offers consistent performance for HPLC systems.
- C18 chemistry for reversed phase separation
- Silica particle substrate with spherical shape
- Low silanol activity and endcapped for stability
- Particle size of 5 μm
- Pore size of 100 Å
- pH range from 2 to 8
- Maximum pressure of 6000 psi (415 Bar)
- Molecular weight range from 1000 to 15000
- Format as a guard column
- Surface area of 300
- Designed for HPLC systems
- USP classification L1
- Inner diameter of 3.9 mm
- Length of 20 mm
- Carbon load of 17%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.