Learn More
Waters Corp Waters Delta-Pak C18 Guard Column, 300Å, 5 μm, 3.9 mm X 20 mm
Catalog No. 50785634 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Delta-Pak C18 Guard Column features stable, bonded, endcapped spherical silica packing with a 5 μm particle size. Designed for reversed-phase separation in HPLC systems, it supports consistent scale-up for milligram-to-gram purification and is primarily utilized for legacy methods.
  • C18 chemistry with spherical silica particles (5 μm).
  • 300 Å pore size; low silanol activity; endcapped.
  • pH range of 2 to 8; max pressure of 6000 psi (415 Bar).
  • Ideal for HPLC systems and legacy method applications.
  • Provides consistent scale-up and easy purity checks of fractions.
  • Dimensions: 3.9 mm inner diameter, 20 mm length.
  • Supplied in a package of 2.
Catalog No. 50-785-634 Supplier Waters Corp Supplier No. WAT046890
The Delta-Pak C18 Guard Column features stable, bonded, endcapped spherical silica packing with a 5 μm particle size. Designed for reversed-phase separation in HPLC systems, it supports consistent scale-up for milligram-to-gram purification and is primarily utilized for legacy methods.
  • C18 chemistry with spherical silica particles (5 μm).
  • 300 Å pore size; low silanol activity; endcapped.
  • pH range of 2 to 8; max pressure of 6000 psi (415 Bar).
  • Ideal for HPLC systems and legacy method applications.
  • Provides consistent scale-up and easy purity checks of fractions.
  • Dimensions: 3.9 mm inner diameter, 20 mm length.
  • Supplied in a package of 2.