Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Guard, C18, 15 μm, 100 Å, 40 mm x 10 mm

Catalog No. 50785443 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Delta-Pak packings separate peptides, proteins, and natural products, using stable, bonded, endcapped spherical silica. Available in C4 or C18 bonded phases and 100Å or 300Å pore sizes, these configurations allow consistent, predictable scale-up purification from milligram to gram scales, with 5μm particles enabling easy purity checks.

  • Designed for the separation of peptides, proteins, and natural products
  • Features C18 chemistry for reversed phase separation
  • Particle size: 15 μm
  • Pore size: 100 Å
  • pH range: 2 to 8
  • Max temperature: 45 °C
  • Max pressure: 1500 psi (100 Bar)
  • Endcapped with low silanol activity
  • Molecular weight range: 1000 to 15000
  • Format: Radial-Pak Guard
  • Dimensions: 40 mm inner diameter x 10 mm length
  • Units per package: 2

Catalog No. 50-785-443 Supplier Waters Corp Supplier No. WAT037842
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delta-Pak packings separate peptides, proteins, and natural products, using stable, bonded, endcapped spherical silica. Available in C4 or C18 bonded phases and 100Å or 300Å pore sizes, these configurations allow consistent, predictable scale-up purification from milligram to gram scales, with 5μm particles enabling easy purity checks.

  • Designed for the separation of peptides, proteins, and natural products
  • Features C18 chemistry for reversed phase separation
  • Particle size: 15 μm
  • Pore size: 100 Å
  • pH range: 2 to 8
  • Max temperature: 45 °C
  • Max pressure: 1500 psi (100 Bar)
  • Endcapped with low silanol activity
  • Molecular weight range: 1000 to 15000
  • Format: Radial-Pak Guard
  • Dimensions: 40 mm inner diameter x 10 mm length
  • Units per package: 2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.