Learn More
Waters Corp Delta-Pak C18 Radial-Pak Guard, 300Å Pore, 15 μm Particle, 40 mm × 10 mm
Catalog No. 50785444 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These packings are designed for the separation of peptides, proteins, and natural products. They are based on a highly stable, bonded, endcapped spherical silica, available in various pore sizes and bonded phases. These configurations offer consistent and predictable scale-up capabilities for purification.
  • Ideal for the separation of peptides, proteins, and natural products
  • Based on highly stable, bonded, endcapped spherical silica
  • Available in two different pore sizes (100Å and 300Å)
  • Available with C4 or C18 bonded phase
  • Offers consistent and predictable scale-up capabilities for milligram-to-gram scale purification
  • Same chemistry available on a high-efficiency 5μm particle for easy purity checks
Catalog No. 50-785-444 Supplier Waters Corp Supplier No. WAT037845
These packings are designed for the separation of peptides, proteins, and natural products. They are based on a highly stable, bonded, endcapped spherical silica, available in various pore sizes and bonded phases. These configurations offer consistent and predictable scale-up capabilities for purification.
  • Ideal for the separation of peptides, proteins, and natural products
  • Based on highly stable, bonded, endcapped spherical silica
  • Available in two different pore sizes (100Å and 300Å)
  • Available with C4 or C18 bonded phase
  • Offers consistent and predictable scale-up capabilities for milligram-to-gram scale purification
  • Same chemistry available on a high-efficiency 5μm particle for easy purity checks