Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Radial-Pak Guard, C4, 15 μm, 300 Å, 25 mm x 10 mm

Catalog No. 50785468 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These packings separate peptides, proteins, and natural products. They utilize stable, bonded, endcapped 5μm or 15μm spherical silica. Available with C4 or C18 bonded phases and 100Å or 300Å pore sizes, they provide consistent scale-up for purification. A 5μm particle option aids purity checks.

  • Chemistry: C4
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Pore size: 300 Å
  • Surface area: 125
  • System: HPLC
  • USP classification: L26
  • Inner diameter: 25 mm
  • Length: 10 mm
  • Carbon load: 3 %
  • Application: Protein
  • Units per package: 2

Catalog No. 50-785-468 Supplier Waters Corp Supplier No. WAT038526
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These packings separate peptides, proteins, and natural products. They utilize stable, bonded, endcapped 5μm or 15μm spherical silica. Available with C4 or C18 bonded phases and 100Å or 300Å pore sizes, they provide consistent scale-up for purification. A 5μm particle option aids purity checks.

  • Chemistry: C4
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Pore size: 300 Å
  • Surface area: 125
  • System: HPLC
  • USP classification: L26
  • Inner diameter: 25 mm
  • Length: 10 mm
  • Carbon load: 3 %
  • Application: Protein
  • Units per package: 2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.