Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, C4, 15 μm, 300 Å, 40 mm × 100 mm

Catalog No. 50785435 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Delta-Pak C4 Radial-Pak Column is designed for the separation of peptides, proteins, and natural products. It features a highly stable, bonded, endcapped 15 μm spherical silica packing with a 300 Å pore size and C4 bonded phase. This column provides consistent and predictable scale-up capabilities for purification.

  • Format: Radial-Pak Column
  • Chemistry: C4
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • pH range: 2–8 pH
  • Maximum pressure: 6000 psi (415 bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1,000 to 30,000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Pore size: 300 Å
  • Inner diameter: 40 mm
  • Length: 100 mm
  • Carbon load: 3%
  • System: HPLC
  • USP classification: L26
  • Application: Protein
  • Units per package: 1

Catalog No. 50-785-435 Supplier Waters Corp Supplier No. WAT037700
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Delta-Pak C4 Radial-Pak Column is designed for the separation of peptides, proteins, and natural products. It features a highly stable, bonded, endcapped 15 μm spherical silica packing with a 300 Å pore size and C4 bonded phase. This column provides consistent and predictable scale-up capabilities for purification.

  • Format: Radial-Pak Column
  • Chemistry: C4
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • pH range: 2–8 pH
  • Maximum pressure: 6000 psi (415 bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1,000 to 30,000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Pore size: 300 Å
  • Inner diameter: 40 mm
  • Length: 100 mm
  • Carbon load: 3%
  • System: HPLC
  • USP classification: L26
  • Application: Protein
  • Units per package: 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.