Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, C4, 15 μm, 100 Å, 8 mm ID x 100 mm Length

Catalog No. 50785226 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Compression Column Segments are plastic compression cartridge segments that insert into radial compression modules (RCM- holder sold separately) for use on liquid chromatography systems.

  • C4 chemistry for effective separation.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Maximum pressure of 6000 psi (415 Bar).
  • Endcapped for improved performance.
  • Molecular weight range of 1000 to 15000.
  • Spherical particle shape.
  • Particle size of 15 μm.
  • Pore size of 100 Å.
  • Radial-Pak column format.
  • Suitable for HPLC systems.
  • 8 mm inner diameter.
  • 100 mm length.
  • 7% carbon load.
  • Designed for protein applications.
  • One unit per package.

Catalog No. 50-785-226 Supplier Waters Corp Supplier No. WAT025848
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compression Column Segments are plastic compression cartridge segments that insert into radial compression modules (RCM- holder sold separately) for use on liquid chromatography systems.

  • C4 chemistry for effective separation.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • pH range of 2 to 8.
  • Maximum pressure of 6000 psi (415 Bar).
  • Endcapped for improved performance.
  • Molecular weight range of 1000 to 15000.
  • Spherical particle shape.
  • Particle size of 15 μm.
  • Pore size of 100 Å.
  • Radial-Pak column format.
  • Suitable for HPLC systems.
  • 8 mm inner diameter.
  • 100 mm length.
  • 7% carbon load.
  • Designed for protein applications.
  • One unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.