Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Radial-Pak Column, C4 Chemistry, 300Å Pore Size, 15 μm Particle Size

Catalog No. 50785458 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Delta-Pak C4 Radial-Pak columns are ideal for efficient separation of peptides, proteins, and natural products. They feature a stable, bonded, endcapped 15μm spherical silica with 300Å pore size and C4 chemistry, enabling consistent scale-up purification from milligram to gram scales.

  • Radial-Pak column for preparative chromatography
  • C4 chemistry, reversed phase separation mode
  • Silica particle substrate, spherical shape
  • 15 μm particle size, 300 Å pore size
  • 25 mm inner diameter, 100 mm length
  • Endcapped, low silanol activity, 3% carbon load
  • PH range: 2 to 8
  • Maximum pressure: 6000 psi (415 bar)
  • Molecular weight range: 1000 to 30000
  • HPLC system compatibility
  • USP classification L26
  • Application: Protein
  • Consistent and predictable scale-up capabilities for milligram to gram purification

Catalog No. 50-785-458 Supplier Waters Corp Supplier No. WAT038509
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delta-Pak C4 Radial-Pak columns are ideal for efficient separation of peptides, proteins, and natural products. They feature a stable, bonded, endcapped 15μm spherical silica with 300Å pore size and C4 chemistry, enabling consistent scale-up purification from milligram to gram scales.

  • Radial-Pak column for preparative chromatography
  • C4 chemistry, reversed phase separation mode
  • Silica particle substrate, spherical shape
  • 15 μm particle size, 300 Å pore size
  • 25 mm inner diameter, 100 mm length
  • Endcapped, low silanol activity, 3% carbon load
  • PH range: 2 to 8
  • Maximum pressure: 6000 psi (415 bar)
  • Molecular weight range: 1000 to 30000
  • HPLC system compatibility
  • USP classification L26
  • Application: Protein
  • Consistent and predictable scale-up capabilities for milligram to gram purification

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.