Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Delta-Pak C4 Prep Column, C4, 15 μm, 30 mm × 300 mm

Catalog No. 50785067 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Delta-Pak C4 Prep Columns are designed for legacy methods and are based on a highly stable, bonded, endcapped 15 μm spherical silica. These columns provide consistent and predictable scale-up capabilities for milligram-to-gram scale purification.

  • C4 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range from 2 pH to 8 pH
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Molecular weight range from 1000 to 15000
  • Spherical particle shape
  • 15 μm particle size
  • 100 Å pore size
  • Prep column format
  • HPLC system compatible
  • 30 mm inner diameter
  • 300 mm length
  • 7% carbon load
  • Suitable for protein applications

Catalog No. 50-785-067 Supplier Waters Corp Supplier No. WAT011810
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Delta-Pak C4 Prep Columns are designed for legacy methods and are based on a highly stable, bonded, endcapped 15 μm spherical silica. These columns provide consistent and predictable scale-up capabilities for milligram-to-gram scale purification.

  • C4 chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range from 2 pH to 8 pH
  • Maximum pressure of 6000 psi (415 Bar)
  • Endcapped
  • Low silanol activity
  • Molecular weight range from 1000 to 15000
  • Spherical particle shape
  • 15 μm particle size
  • 100 Å pore size
  • Prep column format
  • HPLC system compatible
  • 30 mm inner diameter
  • 300 mm length
  • 7% carbon load
  • Suitable for protein applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.