Learn More
Waters Corp DeltaPak C4 Prep Column, 300Å, 15 µm, 19 mm X 300 mm, 1/pk
Catalog No. 50785070 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Delta-Pak packings, ideal for the separation of peptides, proteins, and natural products, are based on a highly stable, bonded, endcapped 5µm or 15µm spherical silica. Delta-Pak is available in two different pore sizes (100Å and 300Å) with either C4 or C18 bonded phase. Delta-Pak packings are available in different cartridge and column configurations, providing consistent and predictable scale-up capabilities for milligram-to-gram scale purification. Also, by having the same chemistry available on a high-efficiency 5µm particle, you can easily check the purity of your fractions.
Catalog No. 50-785-070 Supplier Waters Corp Supplier No. WAT011814
Delta-Pak packings, ideal for the separation of peptides, proteins, and natural products, are based on a highly stable, bonded, endcapped 5µm or 15µm spherical silica. Delta-Pak is available in two different pore sizes (100Å and 300Å) with either C4 or C18 bonded phase. Delta-Pak packings are available in different cartridge and column configurations, providing consistent and predictable scale-up capabilities for milligram-to-gram scale purification. Also, by having the same chemistry available on a high-efficiency 5µm particle, you can easily check the purity of your fractions.