Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Delta-Pak C4 Prep Column, 15 μm, 300 Å, 30 mm × 300 mm

Catalog No. 50785071 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Delta-Pak packings feature highly stable, bonded, endcapped spherical silica, available in 5μm or 15μm particle sizes and 100Å or 300Å pore sizes with C4 or C18 bonded phase. They provide consistent scale-up for milligram-to-gram purification. This product is primarily used for legacy methods.

  • Chemistry: C4
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Pore size: 300 Å
  • Format: Prep column
  • Surface area: 125
  • System: HPLC preparative
  • USP classification: L26
  • Inner diameter: 30 mm
  • Length: 300 mm
  • Carbon load: 3%
  • Application: Protein

Catalog No. 50-785-071 Supplier Waters Corp Supplier No. WAT011815
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Delta-Pak packings feature highly stable, bonded, endcapped spherical silica, available in 5μm or 15μm particle sizes and 100Å or 300Å pore sizes with C4 or C18 bonded phase. They provide consistent scale-up for milligram-to-gram purification. This product is primarily used for legacy methods.

  • Chemistry: C4
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Pore size: 300 Å
  • Format: Prep column
  • Surface area: 125
  • System: HPLC preparative
  • USP classification: L26
  • Inner diameter: 30 mm
  • Length: 300 mm
  • Carbon load: 3%
  • Application: Protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.