Learn More
Waters Corp Waters Corporation Guard Column, C4, 3.9 mm × 20 mm, 5 μm
Supplier:  Waters Corp WAT046885
Encompass_Preferred
The Delta-Pak C4 Guard Column is designed for legacy methods. It features highly stable, bonded, endcapped spherical silica packings, available in 5μm or 15μm particle sizes and 100Å or 300Å pore sizes with C4 or C18 bonded phases. This design supports consistent scale-up for milligram-to-gram purification.
  • Available in C4 or C18 bonded phases for diverse applications.
  • Offered in various cartridge and column configurations.
  • Supports consistent and predictable milligram-to-gram scale purification.
  • Enables easy purity checks of fractions with a high-efficiency 5μm particle.
  • Ideal for separating peptides, proteins, and natural products.
Catalog No. 50-785-633