Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Guard Cartridge, C4, 300Å Pore, 15 μm Particle, 40 mm x 10 mm

Catalog No. 50785445 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Delta-Pak C4 Radial-Pak Guard Cartridge protects analytical and preparative columns for peptide, protein, and natural product separations. It features stable, endcapped 15 μm spherical silica with 300Å pore size, ensuring consistent HPLC performance. Its radial design maximizes column life and minimizes backpressure, aiding purification scale-up.

  • Chemistry: C4
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • pH range: 2 pH to 8 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Pore size: 300 Å
  • Format: Radial-Pak Guard
  • System: HPLC
  • Inner diameter: 40 mm
  • Length: 10 mm
  • Carbon load: 3 %
  • Application: Protein
  • Units per package: 2 pk

Catalog No. 50-785-445 Supplier Waters Corp Supplier No. WAT037851
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Delta-Pak C4 Radial-Pak Guard Cartridge protects analytical and preparative columns for peptide, protein, and natural product separations. It features stable, endcapped 15 μm spherical silica with 300Å pore size, ensuring consistent HPLC performance. Its radial design maximizes column life and minimizes backpressure, aiding purification scale-up.

  • Chemistry: C4
  • Separation mode: Reversed Phase
  • Particle substrate: Silica
  • pH range: 2 pH to 8 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Silanol activity: Low
  • Molecular weight range: 1000 to 30000
  • Particle shape: Spherical
  • Particle size: 15 μm
  • Pore size: 300 Å
  • Format: Radial-Pak Guard
  • System: HPLC
  • Inner diameter: 40 mm
  • Length: 10 mm
  • Carbon load: 3 %
  • Application: Protein
  • Units per package: 2 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.