Learn More
Waters Corp Waters Guard-Pak Insert, Delta-Pak C4, 5 μm, 300 Å, 10/pk
Catalog No. 50785402 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Guard-Pak Insert is designed to protect HPLC columns from particulate and chemical contamination in the mobile phase stream. It helps extend the life and maintain the performance of analytical columns, requiring replacement when contaminants appear or initial backpressure increases significantly.
  • Sorbent is Delta-Pak C4, 300Å
  • Particle size is 5 μm
  • Functions as a guard insert
  • Conforms to USP L26 classification
  • Available in packs of 10
  • Shipped dry and requires conditioning with mobile phase before use
  • Adsorption capacity ranges from 10 mg to 40 mg
  • Pressure limit is 40 Mpa (6000 psi)
  • Packing amount is 50 mg to 100 mg
  • Dead volume ranges from 60 μL to 75 μL
Catalog No. 50-785-402 Supplier Waters Corp Supplier No. WAT036865
The Guard-Pak Insert is designed to protect HPLC columns from particulate and chemical contamination in the mobile phase stream. It helps extend the life and maintain the performance of analytical columns, requiring replacement when contaminants appear or initial backpressure increases significantly.
  • Sorbent is Delta-Pak C4, 300Å
  • Particle size is 5 μm
  • Functions as a guard insert
  • Conforms to USP L26 classification
  • Available in packs of 10
  • Shipped dry and requires conditioning with mobile phase before use
  • Adsorption capacity ranges from 10 mg to 40 mg
  • Pressure limit is 40 Mpa (6000 psi)
  • Packing amount is 50 mg to 100 mg
  • Dead volume ranges from 60 μL to 75 μL