Learn More
MSC Delta Plus Safety Glasses, Clear, Grey Temples, ANSI Z87.1, 10/Box
Catalog No. 5000768511
Encompass
These safety glasses offer a sophisticated sunglass style with all-day comfort. They feature a dual polycarbonate lens with a curved design and non-slip, padded nylon temples, making them suitable for various industrial and public service applications.
  • Dual polycarbonate lens
  • Curved 9 design
  • Non-slip, padded nylon temples
  • Anti-scratch lens coating
  • Clear lens for color perception
  • High performance optical class for less visual fatigue
  • Impact resistant (ANSI Z87.1 certified)
  • Compatible with respiratory masks and helmets
  • Weighs 28 g
  • Packaged in a box of 10
Catalog No. 50-007-68511 Supplier MSC Supplier No. 18030197
These safety glasses offer a sophisticated sunglass style with all-day comfort. They feature a dual polycarbonate lens with a curved design and non-slip, padded nylon temples, making them suitable for various industrial and public service applications.
  • Dual polycarbonate lens
  • Curved 9 design
  • Non-slip, padded nylon temples
  • Anti-scratch lens coating
  • Clear lens for color perception
  • High performance optical class for less visual fatigue
  • Impact resistant (ANSI Z87.1 certified)
  • Compatible with respiratory masks and helmets
  • Weighs 28 g
  • Packaged in a box of 10