Learn More
Delta T™ EDU System
SCI_ED
Catalog No. S96359
Click to view available options
Type:
Delta T EDU System
Catalog No. Type
S96359 Delta T EDU System
1 options
Catalog No. S96359 Supplier Bioptechs Inc Supplier No. 04200504212
Add to Cart
Add to Cart
Includes
  • Controller plus stand
  • Universal stage adapter
  • 2 Culture cylinders
  • 10 Delta T EDU dishes
  • 22mm spacer ring and coverslip weight
Inspire biology students with hands-on, live-cell discoveries in the classroom.
This system makes it easy, safe and convenient to interactively work with live specimens. Discover the excitement of watching microscopic cellular events unfold, while providing a dynamic understanding of biological activity.
  • Accurate and safe temperature control in a high-numeric-aperture compatible dish
  • No hot surfaces; all heat is contained in the dish
  • Thermal regulation via instantaneous low voltage analog control of specially coated dishes
  • Accommodates insect, yeast, mammalian, and avian specimens
  • Compatible with all models of both upright and inverted microscopes
  • Includes a book of protocols and exciting, curriculum relevant demonstrations
Specifications
Type Delta T EDU System
For Use With (Equipment) Models of both upright and inverted microscope
Includes Controller plus stand, universal stage adapter, 2 culture cylinders, 10 delta T EDU dishes, 22mm spacer ring and coverslip weight