Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Delta-tocotrienol | 25612-59-3 | 98.1% | 10 MG
SDP

Catalog No. 5000230743
Encompass_Preferred

Delta-tocotrienol is a vitamin E family compound used as an analytical standard and research reagent with antioxidant and lipid-modulating activity. It is supplied as a high-purity reference material for laboratory assays.

  • CAS number 25612-59-3.
  • IUPAC name (2R)-2,8-dimethyl-2-[(3E,7E)-4,8,12-trimethyltrideca-3,7,11-trienyl]-3,4-dihydrochromen-6-ol.
  • Molecular formula C27H40O2.
  • Molecular weight 396.6 g/mol.
  • Purity 98.1%.
  • Provided as a 10 mg solid vial for analytical use.

Catalog No. 50-002-30743 Supplier Medchemexpress LLC Supplier No. HY12277810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delta-tocotrienol is a vitamin E family compound used as an analytical standard and research reagent with antioxidant and lipid-modulating activity. It is supplied as a high-purity reference material for laboratory assays.

  • CAS number 25612-59-3.
  • IUPAC name (2R)-2,8-dimethyl-2-[(3E,7E)-4,8,12-trimethyltrideca-3,7,11-trienyl]-3,4-dihydrochromen-6-ol.
  • Molecular formula C27H40O2.
  • Molecular weight 396.6 g/mol.
  • Purity 98.1%.
  • Provided as a 10 mg solid vial for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.