Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Undecanolactone 98.0+%, TCI America™
SDP

Catalog No. H034425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H034425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H034425G Supplier TCI America Supplier No. H034425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 710-04-3
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00006650
InChI Key YZRXRLLRSPQHDK-UHFFFAOYSA-N
Synonym 5-Hydroxyundecanoic Acid delta-Lactone, delta-Undecalactone
PubChem CID 61204
IUPAC Name 6-hexyloxan-2-one
SMILES CCCCCCC1CCCC(=O)O1

Specifications

Color Yellow
Boiling Point 168°C
Quantity 25 g
Formula Weight 184.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material delta-Undecanolactone
EINECSNumber (5)-0698&(9)-0137
RTECSNumber UQ1320000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.