Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DELTA9(11)-Methyltestosterone, TCI America™
SDP

Catalog No. M0638100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0638100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0638100MG Supplier TCI America Supplier No. M0638100MG

Chemical Identifiers

CAS 1039-17-4
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.442
MDL Number MFCD00021151
InChI Key BFMZZSBHQOCQFQ-HTDHLNIYSA-N
Synonym 17beta-Hydroxy-17-methylandrosta-4,9(11)-dien-3-one
PubChem CID 220507
ChEBI CHEBI:34182
IUPAC Name (8S,10S,13S,14S,17S)-17-hydroxy-10,13,17-trimethyl-2,6,7,8,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-3-one
SMILES CC12CCC(=O)C=C1CCC3C2=CCC4(C3CCC4(C)O)C

Specifications

Melting Point 169°C
Quantity 100 mg
Formula Weight 300.44
Percent Purity ≥90.0% (GC)
Physical Form Granules
Chemical Name or Material DELTA9(11)-Methyltestosterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.