Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deltamethric acid | 53179-78-5 | 99.8% | 297.97 g/mol | C8H10Br2O2 | 10 MG
SDP

Catalog No. 5000284178
Encompass_Preferred

Deltamethric acid is a chemical intermediate used in the synthesis of the pyrethroid insecticide deltamethrin. It is supplied for laboratory research and development purposes and is not intended for human or veterinary use. Refer to the product datasheet and SDS for batch-specific handling, storage, and safety information.

  • Used as an intermediate in deltamethrin synthesis.
  • Intended for research use only; not for human consumption.
  • High purity: 99.8% (HPLC).
  • Molecular formula C8H10Br2O2; molecular weight 297.97 g/mol.
  • CAS number 53179-78-5 for unambiguous identification.
  • Typically supplied in small milligram quantities (e.g., 1-100 mg) suitable for laboratory synthesis.
  • Consult the safety data sheet for handling, storage, and disposal instructions.

Catalog No. 50-002-84178 Supplier Medchemexpress LLC Supplier No. HYW51711010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Deltamethric acid is a chemical intermediate used in the synthesis of the pyrethroid insecticide deltamethrin. It is supplied for laboratory research and development purposes and is not intended for human or veterinary use. Refer to the product datasheet and SDS for batch-specific handling, storage, and safety information.

  • Used as an intermediate in deltamethrin synthesis.
  • Intended for research use only; not for human consumption.
  • High purity: 99.8% (HPLC).
  • Molecular formula C8H10Br2O2; molecular weight 297.97 g/mol.
  • CAS number 53179-78-5 for unambiguous identification.
  • Typically supplied in small milligram quantities (e.g., 1-100 mg) suitable for laboratory synthesis.
  • Consult the safety data sheet for handling, storage, and disposal instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.