Learn More
Cayman Chemical Deltamethrin, Purity: ≥95%, CAS: 52918-63-5, 50 mg
SDP
Catalog No. 502936893
Encompass_Preferred
A type II pyrethroid insecticide and Nav channel modulator; activates Nav1.8 channels and induces a tail current delay in Xenopus oocytes expressing the rat channel at 10 µM; induces mortality in A. sinensis (LC50 = 4.522 ppm) and T. urticae (LC50 = 0.06 g/L); decreases sperm count, motility, and viability and increases the percentage of morphologically abnormal sperm in rats at 5 mg/kg
Catalog No. 50-293-6893 Supplier Cayman Chemical Supplier No. 2417250 MG
A type II pyrethroid insecticide and Nav channel modulator; activates Nav1.8 channels and induces a tail current delay in Xenopus oocytes expressing the rat channel at 10 µM; induces mortality in A. sinensis (LC50 = 4.522 ppm) and T. urticae (LC50 = 0.06 g/L); decreases sperm count, motility, and viability and increases the percentage of morphologically abnormal sperm in rats at 5 mg/kg