Learn More
Medchemexpress LLC Deltamethrin | 52918-63-5 | 250 MG
SDP
Catalog No. 5000413958
Encompass_Preferred
Deltamethrin (Decamethrin) is an orally active synthetic pyrethroid insecticide. It induces oxidative stress and results in inflammation and apoptosis by inhibiting the Nrf2/HO-1 pathway. Deltamethrin also exhibits an anticancer effect by inducing apoptosis and can be used extensively in pest control. This product is an analytical standard, intended for research and analytical applications.
  • Orally active synthetic pyrethroid insecticide
  • Induces oxidative stress
  • Results in inflammation and apoptosis via inhibiting Nrf2/HO-1 pathway
  • Has an anticancer effect by inducing apoptosis
  • Can be used extensively in pest control
  • Relevant to research in neurological disease, inflammation/immunology, and cancer
  • Analytical standard for research and analytical applications
Catalog No. 50-004-13958 Supplier Medchemexpress LLC Supplier No. 250MGHYB1971
Deltamethrin (Decamethrin) is an orally active synthetic pyrethroid insecticide. It induces oxidative stress and results in inflammation and apoptosis by inhibiting the Nrf2/HO-1 pathway. Deltamethrin also exhibits an anticancer effect by inducing apoptosis and can be used extensively in pest control. This product is an analytical standard, intended for research and analytical applications.
  • Orally active synthetic pyrethroid insecticide
  • Induces oxidative stress
  • Results in inflammation and apoptosis via inhibiting Nrf2/HO-1 pathway
  • Has an anticancer effect by inducing apoptosis
  • Can be used extensively in pest control
  • Relevant to research in neurological disease, inflammation/immunology, and cancer
  • Analytical standard for research and analytical applications