Learn More
Braintree Scientific DELTAPHASE ISOTHERMAL PAD 3SET
SDP
Catalog No. NC9411230
Encompass
The Deltaphase Isothermal Pad acts as a source of heat, but cannot overheat. Its constant temperature is based on a thermodynamic principle and needs no electricity or wires, no thermostat or feedback **Sold for research purposes only and not for human use**
Catalog No. NC9411230 Supplier Braintree Scientific Supplier No. 39DP
The Deltaphase Isothermal Pad acts as a source of heat, but cannot overheat. Its constant temperature is based on a thermodynamic principle and needs no electricity or wires, no thermostat or feedback **Sold for research purposes only and not for human use**