Learn More
Braintree Scientific Isothermal Pads 6"" x 4""
SDP
Catalog No. 501954665
Encompass
Isothermal Pad is a new approach to body temperature stabilization. It acts as a source of heat, but cannot overheat once it has reached it's phase change . Its constant temperature is based on a thermodynamic principle and needs no electricity or wires, no thermostat or feedback controller. It cannot generate electrical signals or cause interference. It cannot trigger an explosion. It can maintain a small animal at near normal and constant temperature for several hours. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4665 Supplier Braintree Scientific Supplier No. DPSASB
Isothermal Pad is a new approach to body temperature stabilization. It acts as a source of heat, but cannot overheat once it has reached it's phase change . Its constant temperature is based on a thermodynamic principle and needs no electricity or wires, no thermostat or feedback controller. It cannot generate electrical signals or cause interference. It cannot trigger an explosion. It can maintain a small animal at near normal and constant temperature for several hours. **Sold for research purposes only and not for human use**