Learn More
Braintree Scientific DELTAPHASE OP BOARD/2 PADS
SDP
Catalog No. NC9877276
Encompass
This compact operating board takes advantage of the Deltaphase principle to maintain animal body temperature during surgical or experimental procedures. The unit is supplied with 2 Deltaphase pads. **Sold for research purposes only and not for human use**
Catalog No. NC9877276 Supplier Braintree Scientific Supplier No. 39OP
This compact operating board takes advantage of the Deltaphase principle to maintain animal body temperature during surgical or experimental procedures. The unit is supplied with 2 Deltaphase pads. **Sold for research purposes only and not for human use**