Learn More
Medchemexpress LLC HY-15747 5mg Medchemexpress, Deltarasin CAS:1440898-61-2 Purity:>98%
SDP
Catalog No. 502081789
Encompass_Preferred
Medchemexpress, HY-15747 5mg Deltarasin CAS:1440898-61-2 Deltarasin is an inhibitor of KRAS-PDEδ interaction with Kd of 38 nM for binding to purified PDEδ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1789 Supplier Medchemexpress LLC Supplier No. HY157475MG
Medchemexpress, HY-15747 5mg Deltarasin CAS:1440898-61-2 Deltarasin is an inhibitor of KRAS-PDEδ interaction with Kd of 38 nM for binding to purified PDEδ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.