Learn More
Medchemexpress LLC HY-15747A 5mg Medchemexpress, Deltarasin (hydrochloride) CAS:1613404-76-4 Purity:>98%
SDP
Catalog No. 502081790
Encompass_Preferred
Medchemexpress, HY-15747A 5mg Deltarasin (hydrochloride) CAS:1613404-76-4 Deltarasin hydrochloride is an inhibitor of KRAS-PDEδinteraction with Kd of 38 nM for binding to purified PDEδ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1790 Supplier Medchemexpress LLC Supplier No. HY15747A5MG
Medchemexpress, HY-15747A 5mg Deltarasin (hydrochloride) CAS:1613404-76-4 Deltarasin hydrochloride is an inhibitor of KRAS-PDEδinteraction with Kd of 38 nM for binding to purified PDEδ. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.