Learn More
Cayman Chemical DeltarasInhydrochlorIde 10mg
SDP
Catalog No. 502892644
Encompass_Preferred
A small molecule that binds to the farnesyl-binding pocket of PDEδ (Kd = 41 nM) in cells thus inhibiting its interaction with KRAS and disrupting RAS signaling; reduces proliferation and increases cell death of Panc-Tu-I tumor cells at 5 μM in a human pancreatic ductal adenocarcinoma cell model; reduces tumor growth at a dose of 10 mg/kg in nude mice xenografted with pancreatic carcinoma
Catalog No. 50-289-2644 Supplier Cayman Chemical Supplier No. 900153610 MG
A small molecule that binds to the farnesyl-binding pocket of PDEδ (Kd = 41 nM) in cells thus inhibiting its interaction with KRAS and disrupting RAS signaling; reduces proliferation and increases cell death of Panc-Tu-I tumor cells at 5 μM in a human pancreatic ductal adenocarcinoma cell model; reduces tumor growth at a dose of 10 mg/kg in nude mice xenografted with pancreatic carcinoma