Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Deltasonamide 2 hydrochloride | 2448341-55-5 | 99.8% | 5 MG
SDP

Catalog No. 5000231294
Encompass_Preferred

Deltasonamide 2 hydrochloride is the hydrochloride salt of Deltasonamide 2, a competitive, high-affinity inhibitor of PDEδ (phosphodiesterase delta) with a reported Kd of ~385 pM. It is supplied as a solid research reagent, soluble in DMSO and water, and intended for biochemical and cellular studies probing PDEδ function.

  • High reported binding affinity to PDEδ (Kd ~385 pM).
  • High purity (reported 99.78%).
  • Molecular weight 683.71 g/mol and formula C30H40Cl2N6O4S2.
  • Hydrochloride salt form provided as a solid.
  • Soluble in DMSO and water for assay preparation.
  • Suitable for biochemical and cellular research applications.

Catalog No. 50-002-31294 Supplier Medchemexpress LLC Supplier No. HY122641C5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deltasonamide 2 hydrochloride is the hydrochloride salt of Deltasonamide 2, a competitive, high-affinity inhibitor of PDEδ (phosphodiesterase delta) with a reported Kd of ~385 pM. It is supplied as a solid research reagent, soluble in DMSO and water, and intended for biochemical and cellular studies probing PDEδ function.

  • High reported binding affinity to PDEδ (Kd ~385 pM).
  • High purity (reported 99.78%).
  • Molecular weight 683.71 g/mol and formula C30H40Cl2N6O4S2.
  • Hydrochloride salt form provided as a solid.
  • Soluble in DMSO and water for assay preparation.
  • Suitable for biochemical and cellular research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.